N-(5-(4-Hydroxyphenyl)thiazol-2-yl)acetamide,95% 1 * 0,1 g
Catalog # AMBDA1107429-100MG
Supplier: AMBEED
N-(5-(4-Hydroxyphenyl)thiazol-2-yl)acetamide,95% 1 * 0,1 g
Catalog # AMBDA1107429-100MG
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Pyridine ≥99%, TECHNICAL
Supplier: VWR Chemicals
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific