Potassium (Z)-(2,3-dibromoprop-1-en-1-yl)trifluoroborate,95% mix TBC as stabilizer 1 * 0,1 g
Catalog # AMBDA986293-100MG
Supplier: AMBEED
Potassium (Z)-(2,3-dibromoprop-1-en-1-yl)trifluoroborate,95% mix TBC as stabilizer 1 * 0,1 g
Catalog # AMBDA986293-100MG
Supplier: AMBEED
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals