3-Methyloxolan-3-amine hydrochloride,98% 1 * 0,1 g
Catalog # AMBDA845399-100MG
Supplier: AMBEED
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
Petroleum spirit, 40…60 °C ACS
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.7% (by GC), CHROMASOLV™ for pesticide residue analysis acc. to FDA, Riedel-de Haën™
Supplier: Honeywell Chemicals