(1S,2S)-2-(4-Fluorophenyl)cyclopropanecarboxylic acid,95% 1 * 1 g
AMBDA814958-1G
: AMBEED
Frequently Bought Together
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
1,4-Dioxane ≥99% for HPLC
: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
: Honeywell Chemicals
Cyclohexanone ≥99.0% analytical reagent
: VWR Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
Xylene - Mixture of isomers technical grade 1 * 5 L
: PanReac AppliChem
Bromoform 99+% stabilised
: Thermo Fisher Scientific