(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol,98% 1 * 0,5 g
Catalog # AMBDA520898-50MG
Supplier: AMBEED
(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol,98% 1 * 0,5 g
Catalog # AMBDA520898-50MG
Supplier: AMBEED
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol eurodenatured ≥80%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific