2,5-Dimethoxypyridine,95% 1 * 0,25 g
Catalog # AMBDA311075-250MG
Supplier: AMBEED
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone, EMPLURA®, Reagent Grade for synthesis, Supelco®
Supplier: Merck
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-ETHOXYETHANOL, REAGENTPLUS, 99%. 1 * 1 Each
Supplier: Merck
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific