Cold trap, type S
Cold finger, S 29-Z, borosilicate glass 3,3, spherical joint S29, 150 ml condensate capacity and length of 200 mm
Artikelnr. KGWA17405
Anbieter: KGW
Jump to:
Product Details & Documents
Type S cold trap has a conical ground joint NS 45/40 as connection part. The condensation area can be separated from the upper section of cold trap. Therefore it is easy to pour out the condensate and to clean the whole cold trap. The connection part NS 45/40 is secured by a spring clamp out of steel.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
Specifications
- Description:Cold finger, S 29-Z, borosilicate glass 3,3, spherical joint S29, 150 ml condensate capacity and length of 200 mm
- Seq:01