[EN]MYCOPLASMA GALLISEPTICUM PCR STANDAR 1 * 1 ST
Catalog # MSPP52-0115
Supplier: MARKET SOURCE PART PROCESS
[EN]MYCOPLASMA GALLISEPTICUM PCR STANDAR 1 * 1 ST
Catalog # MSPP52-0115
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Refractive index standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Alizarin red S (sodium salt) solution Reag. Ph. Eur. 1002601
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
Buffers, concentrated solutions, Titrisol®, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach