VWR®, Cryo coder cap inserts
Cap insert, Yellow
Supplier: VWR Collection
Jump to:
Product Details & Documents
Product can be used as coding/identification of samples types stored in it.
Cryocoder/cap inserts are available in multiple colour options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).
Specifications
- Colour:Yellow
- Description:Cap insert
- Seq:12