Regulated BlueNotes 420 is designed to provide the features required to enable users who need to do so, to operate Sherwood Dual Channel Flame Photometers in a way, compliant with data handling standard 21 CFR Part 11. 1 * 1 items
Catalog # SHEW42066100
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Frequently Bought Together