Service Kit, Power Board Model(s): SU780XLE Serial Range: 19070-10069 to Present 1 * 1 KIT
Catalog # GLCLSKSU780XLE7500
Supplier: Stirling Ultracold
Service Kit, Power Board Model(s): SU780XLE Serial Range: 19070-10069 to Present 1 * 1 KIT
Catalog # GLCLSKSU780XLE7500
Supplier: Stirling Ultracold
Frequently Bought Together
Accessories for accu-jet® pro
Supplier: Brand
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Service Kit, Inverter PCB Model(s): SU780XLE Serial Range: 19070-10069 to Present 1 * 1 KIT
Supplier: Stirling Ultracold
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG