SEAL FOR 6-WAY VALVE 404 1 * 1 items
Catalog # GILIF401504010
Supplier: GILSON LCMS
Frequently Bought Together
Guillotine tubing cutter
Supplier: VICI JOUR RESEARCH
Gas detection tubes for nitrous fumes
Supplier: Dräger
Gas detection tube for hydrogen sulphide
Supplier: Dräger
Accessories for titrators
Supplier: Mettler - Toledo
VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
Supplier: VWR Collection
Quick connect Luer adapters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Flow indicator paddle wheel
Supplier: Brand
OIL IMPACTOR 0.1-1.0 MG/M3 1 * 10 items
Supplier: Dräger
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Bellows pumps, accuro®
Supplier: Dräger
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
Supplier: Mettler - Toledo
Multi-parameter meters, bench, HQd (HQ430d / HQ440d)
Supplier: Hach
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
Multi-parameter testers, Pocket Pro™+
Supplier: Hach
Accessories for buchi melting point apparatus M560 and M565
Supplier: BUCHI