ContactUsLinkComponent
Light source with two self-sustaining gooseneck-type guides equipped with 2x 3 Watt white 6500 K power LED. Each self-sustaining Gooseneck-type LED guide is equipped with a 3-lens focusing head and has a length of 56 cm. Luminous flux 25,000 lux Oper 1 * 1 ST
Catalog # EUMXLE5207
Supplier:  EUROMEX
Light source with two self-sustaining gooseneck-type guides equipped with 2x 3 Watt white 6500 K power LED. Each self-sustaining Gooseneck-type LED guide is equipped with a 3-lens focusing head and has a length of 56 cm. Luminous flux 25,000 lux Oper 1 * 1 ST
Catalog # EUMXLE5207
Supplier:  EUROMEX
Frequently Bought Together