SUCCINYLSULFATHIAZOLE 1 * 100 g
SPCMS2591-100GM
Frequently Bought Together
Sodium hydroxide 35% in aqueous solution, TECHNICAL
: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
: VWR Chemicals
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Accessories
: Hach
pH indicator test strips, pH-Fix
: MACHEREY-NAGEL