TETRAMETHYLAMMONIUM BROMIDE REAGENT ACS 1 * 500 g
Catalog # SPCMT2577-500GM
Supplier: Spectrum Chemical
TETRAMETHYLAMMONIUM BROMIDE REAGENT ACS 1 * 500 g
Catalog # SPCMT2577-500GM
Supplier: Spectrum Chemical
Frequently Bought Together
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Lithium perchlorate trihydrate 99%, extra pure
Supplier: Thermo Fisher Scientific
Liquid for manometer with sloping tube
Supplier: VWR Chemicals
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
(±)-2-Methyltetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl