ContactUsLinkComponent
RETSCH Test Sieve ASTM E 118x2 500µm 35Mesh stainless steel with calibration certificate acc. EN 10204 3.1 1 * 1 items
Catalog # RETS60.350.000500
Supplier:  Retsch GmbH
RETSCH Test Sieve ASTM E 118x2 500µm 35Mesh stainless steel with calibration certificate acc. EN 10204 3.1 1 * 1 items
Catalog # RETS60.350.000500
Supplier:  Retsch GmbH
Frequently Bought Together