alkaline, foam-inhibited cleaning concentrate elma clean 275 dip/splash 1 * 25 L
Catalog # ELMA8000082
Supplier: ELMA SCHMIDBAUER
alkaline, foam-inhibited cleaning concentrate elma clean 275 dip/splash 1 * 25 L
Catalog # ELMA8000082
Supplier: ELMA SCHMIDBAUER
Frequently Bought Together
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Butyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, Sigma-Aldrich®
Supplier: Merck
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific