ContactUsLinkComponent
External Dimensions: 565 x 510 x 435 mm Internal Dimensions: 444 x 388 x 302 mm Dry-Ice Mass: 33.1 kg Payload Volume: 16.8 L Duration: 96+ 1 * 1 items
Catalog # DGINFRAPHT45
Supplier:  DGP INTELSIUS
External Dimensions: 565 x 510 x 435 mm Internal Dimensions: 444 x 388 x 302 mm Dry-Ice Mass: 33.1 kg Payload Volume: 16.8 L Duration: 96+ 1 * 1 items
Catalog # DGINFRAPHT45
Supplier:  DGP INTELSIUS
Frequently Bought Together