THERMAL PAPER ROLL 58MM DIAM 50MM 1 * 10 items
Catalog # METT30094723
Supplier: Mettler - Toledo
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone, EMPLURA®, Reagent Grade for synthesis, Supelco®
Supplier: Merck
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Potassium chloride 3 mol/l (3 N) in aqueous solution
Supplier: Mettler - Toledo
Disposable cuvettes
Supplier: Brand