STAY SAFE PURGING BOTTLE 1 * 1 items
HEWL5043-1339
: Agilent
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
Hydra square bottle, GL 45, with 4 side necks, DURAN®
: DWK Life Sciences
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Sodium azide ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Accessories for cell counter, EVE™
: NANOENTEK
Gold standard syringes for autosampler
: Agilent
Sodium-1-decanesulphonate, anhydrous 99+% for ion pair chromatography
: Thermo Fisher Scientific
Propionyl chloride ≥98%
: Thermo Fisher Scientific