Containers with Sloping Base
Hygienic valve kit with EPDM seals, —
Catalog # SAMPA136-01A
Supplier: Sampling Systems
Springe zu:
Product Details & Documents
These drums which incorporate a sloping base allow the container to drain completely.
- Made from 316L stainless steel
- Includes lid, clamp and silicone seal
- Other seals available
- Ferrule for easy emptying
- Handles enable easy transportation
The Sampling Systems range of containers with sloping base ensures that material can be completely drained from within and include a ferrule at the base for easy emptying. They have a mirror polish finish for easy cleaning.
Spezifikationen
- Capacity:—
- Description:Hygienic valve kit with EPDM seals
- Seq:05