ContactUsLinkComponent
Guardian 3000 Hotplate-Stirrer e-G31HS04C 230V EU, Temperature Range: Ambient + 5°C – 500°C, Capacity 15L, Top Plate Dimensions: 10,2 x 10,2 cm, Ceramic 1 * 1 items
Catalog # OHAU30680280
Supplier:  OHAUS
Guardian 3000 Hotplate-Stirrer e-G31HS04C 230V EU, Temperature Range: Ambient + 5°C – 500°C, Capacity 15L, Top Plate Dimensions: 10,2 x 10,2 cm, Ceramic 1 * 1 items
Catalog # OHAU30680280
Supplier:  OHAUS
Frequently Bought Together