ContactUsLinkComponent
TMFC Bin SA44. Maximum Flow Rate (sccm) for Air Sparge, O2, N2 & Air Overlay is 302 - 926 and for CO2 is 226 - 691. Operating Range is 2% of Desired Maximum Flow Rate 1 * 1 items
Catalog # DISI5000-2001
Supplier:  DISTEK
TMFC Bin SA44. Maximum Flow Rate (sccm) for Air Sparge, O2, N2 & Air Overlay is 302 - 926 and for CO2 is 226 - 691. Operating Range is 2% of Desired Maximum Flow Rate 1 * 1 items
Catalog # DISI5000-2001
Supplier:  DISTEK
Frequently Bought Together