Order Entry
Austria
Orders LinkContactUsLinkComponent
Elemental Impurities Mixture, Reference Standards

Elemental Impurities Mixture, Reference Standards

Elemental Impurities Class 2B Mixture, USP reference standard, Bottle, 120 mL
Artikelnr. 1234427.
Anbieter:   USP
Elemental Impurities Mixture, Reference Standards
Artikelnr. 1234427.
Anbieter:  USP
CAS Nummer:  
Elemental Impurities Mixture, Reference Standards

Elemental Impurities Class 2B Mixture, USP reference standard, Bottle, 120 mL

CAS Nummer:  
Preis
576,00 €
Log in for availability
1 * 120 mL
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    120 mL
  • Standards:
    USP reference standard
  • Description:
    Elemental Impurities Class 2B Mixture
  • UN No.:
    1789
  • Cat. No.:
    1234427.
  • Description:
    Elemental Impurities Class 2B Mixture

Specifications