Order Entry
Austria
Orders LinkContactUsLinkComponent
RIPA Buffer, Abcam

RIPA Buffer, Abcam

RIPA Buffer
Catalog # ABCAAB288006-500
Marketsource
Supplier:   Abcam
RIPA Buffer, Abcam
Catalog # ABCAAB288006-500
Supplier:  Abcam
Marketsource
CAS Number:  
RIPA Buffer, Abcam

RIPA Buffer

CAS Number:  
Price
436,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

A ready-to-use buffer useful for lysing cells and tissues for radioimmunoprecipitation assay.

  • Constituents: 1% Alcohol Ethoxylate, 0.877% Sodium chloride, 0.788% Tris HCl, 0.5% Sodium deoxycholate, 0.1% Sodium lauryl sulfate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    RIPA Buffer
  • Cat. No.:
    ABCAAB288006-500
  • Storage temperature:
    Store at -20°C.

Specifications