Sterile TechniCloth STX4351, cleanroom dry sterile nonwoven wipe, 45% polyester / 55% cellulose, 9in x 11in (23x28cm), SAL 10
Catalog # TEXWSTX4351
Supplier: Texwipe
Sterile TechniCloth STX4351, cleanroom dry sterile nonwoven wipe, 45% polyester / 55% cellulose, 9in x 11in (23x28cm), SAL 10
Catalog # TEXWSTX4351
Supplier: Texwipe
Frequently Bought Together
Hand Towels, KLEENEX® Ultra Slimroll™
Supplier: KIMBERLY CLARK
Cleanroom wipes, Anticon® Gold Sorb™
Supplier: CONTEC
Accessories for pH/°C meter, handheld, 230
Supplier: Testo
AutoFoam Dispensers
Supplier: Rubbermaid Commercial Products
Accessories for YSI meters
Supplier: YSI ENVIRONMENTAL
Electrode Cables and Accessories
Supplier: CONSORT
pH combination electrodes, IDS®
Supplier: SI Analytics
Optimised Filling Tools for MAS Rotors
Supplier: MARKET SOURCE PART PROCESS
pH electrodes for Knick® Portamess® meters
Supplier: HAMILTON BONADUZ
pH electrodes, Eutech
Supplier: EUTECH