Sample ring kit model A, standard set to a depth of 2 m. For soil sample rings O 53 mm. Soil sample ring kits are used for a variety of tests, including bulk density determination of soil. 1 * 1 SET
Catalog # EIJK0753SA
Supplier: EIJKELKAMP AGRISECTOR
Frequently Bought Together