SENSITIVITY STANDARDS-MYCOPLASMA10CFU 1 * 1 ST
Catalog # MSPP102-1103
Supplier: MARKET SOURCE PART PROCESS
SENSITIVITY STANDARDS-MYCOPLASMA10CFU 1 * 1 ST
Catalog # MSPP102-1103
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
p-Xylene-[D₁₀]
Supplier: EHRENSTORFER
Milbemectin A4 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
Quizalofop (free acid) D3 (3,3,3-D3) 100 ng/µl 1 * 1,1 mL
Supplier: EHRENSTORFER
Acetaldehyde, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, Oleanolic Acid
Supplier: Merck
Organic reference standards, Tramadol, Ph.Eur., RS
Supplier: EDQM
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Microcystin RR-YR-LR solution, analytical standard, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, F.A.M.E. Mix RM-4
Supplier: Merck
Sodium standard solution, 1000 mg/L Na in water standard for ICP
Supplier: CPACHEM
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Terbumeton certified 1 * 0,25 g
Supplier: EHRENSTORFER