Reagecon pH 8.00 Buffer Solution at 38°C 1 * 1 L
Catalog # REGE108038
Supplier: REAGECON
Frequently Bought Together
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Accessories for pH/°C meter, handheld
Supplier: Testo
Calcium 1.000 mg/l in water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Buffer solutions
Supplier: REAGECON
Conductivity standards
Supplier: HAMILTON BONADUZ
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond