BRAND® BIO-CERT® Robotic Tips, PP
Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY, 40 - 1000 µl, -, -, 1 pack (960 pieces)
Catalog # BRND734152
Supplier: Brand
Jump to:
Product Details & Documents
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses with the pietting robot liquid handling station.
- Raw materials are free from DiHEMDA und Oleamid
- Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged
- No parting agents or demolding aids are used
- BIO-CERT® Liquid handling quality: Free of human DNA, RNase, DNase, pyrogens, PCR-inhibitors and ATP
For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the liquid handling station pipetting robot.
Specifications
- Packed:1 pack (960 pieces)
- Sterile:-
- Volume:40 - 1000 µl
- Seq:13
- Tip style:Racked, TipRack (refill system), BIO-CERT® LIQUID HANDLING QUALITY
- Filter:-