ContactUsLinkComponent
B1-220E-SP (4X, 10X, 40X) Binocular head, 45º inclined, 360º rotating Widefield eyepieces WF10X/18mm Quadruple revolving nosepiece Semi-Plan objectives SP 4X 10X ,40X S Coaxial coarse and fine focusing system with tension adjustment Built-in low pos 1 * 1 items
Catalog # MOTI1100100501157
Supplier:  MOTIC
B1-220E-SP (4X, 10X, 40X) Binocular head, 45º inclined, 360º rotating Widefield eyepieces WF10X/18mm Quadruple revolving nosepiece Semi-Plan objectives SP 4X 10X ,40X S Coaxial coarse and fine focusing system with tension adjustment Built-in low pos 1 * 1 items
Catalog # MOTI1100100501157
Supplier:  MOTIC
Frequently Bought Together