ContactUsLinkComponent
Masterflex® Gold-colored IDentifier Hinge Clamp, 2-1/2" Clamp Size, 304 SS; 1/EA
Masterflex® Gold-colored IDentifier Hinge Clamp, 2-1/2" Clamp Size, 304 SS; 1/EA
Artikelnr. MFLX30562-54
undefined
Masterflex® Gold-colored IDentifier Hinge Clamp, 2-1/2" Clamp Size, 304 SS; 1/EA
Artikelnr. MFLX30562-54
About this item
Easy visual identification of critical joints for review in your process.
  • Constructed of gold-coloured 304 Stainless Steel
Easily distinguishable from standard stainless steel clamps from a distance, these gold-coloured sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.
  • Connection type:
    Sanitary
  • Description:
    Gold-coloured Identifier Hinge Clamp, 2-1/2" Clamp Size, 304 SS; 1/EA
  • Material:
    Stainless steel 304
  • Environmentally Preferable:
  • Sanitary clamp size (port 1):
    2"
  • Warranty:
    1 year
  • Pk:
    1
  • Clamp type:
    Sanitary
Frequently Bought Together