ContactUsLinkComponent
Masterflex® DB15 Male Connector; for Creating Your Own Cable
Masterflex® DB15 Male Connector; for Creating Your Own Cable
Catalog # MFLX07595-52
undefined
Masterflex® DB15 Male Connector; for Creating Your Own Cable
Catalog # MFLX07595-52
About this item
Create your own cable for optimization of your Masterflex® drive.
  • Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
  • Description:
    DB15 male connector for creating cable
  • Environmentally Preferable:
  • Model:
    07595-52
  • Pk:
    1
Frequently Bought Together