ContactUsLinkComponent
Smart Gasket Sanitary Sampling Gasket, Viton, 3 Port, 1-1/2" Tri-Clamp
Smart Gasket Sanitary Sampling Gasket, Viton, 3 Port, 1-1/2" Tri-Clamp
Catalog # MFLX30544-14
undefined
Smart Gasket Sanitary Sampling Gasket, Viton, 3 Port, 1-1/2" Tri-Clamp
Catalog # MFLX30544-14
About this item
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
  • Sanitary clamp size (Port 1):
    1-1/2"
  • Connection type:
    Sanitary
  • Description:
    1 1/2" Tri-Clamp, 3 Port, Viton, Each
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1
  • Fitting material:
    Viton
  • Sanitary gasket size:
    1-1/2"
  • No. of ports:
    3
  • Warranty:
    1 year
Frequently Bought Together