Micropump L22059 Gear Pump Service Kit for 07003-02; 1/Ea
Catalog # MFLX07003-12
Supplier: Avantor Fluid Handling
About this item
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
- Description:Gear Pump Service Kit, for 07003-02
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
Frequently Bought Together