Order Entry
Austria
ContactUsLinkComponent
Masterflex® Extension Cable for L/S® and I/P® Digital Modular Drives; 9 ft
Masterflex® Extension Cable for L/S® and I/P® Digital Modular Drives; 9 ft
Catalog # MFLX77301-82
undefined
Masterflex® Extension Cable for L/S® and I/P® Digital Modular Drives; 9 ft
Catalog # MFLX77301-82

About this item

Increase modular drive flexibility with extended cable lengths.

  • Easily connect to your Masterflex® pump with connectors on both ends

Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller by 9 ft (2,7 m) to increase installation flexibility. Do not extend more than 25 ft.

  • Description:
    Extension cable for L/S and I/P digital modular drives
  • Length:
    9 ft.
  • Environmentally Preferable:
  • Model:
    77301-82
  • Warranty:
    2 years
  • Pk:
    9 Feet
Frequently Bought Together