ContactUsLinkComponent
Masterflex® L/S® Gear Service Kits for 10 to 600 rpm Digital Modular Drives
Masterflex® L/S® Gear Service Kits for 10 to 600 rpm Digital Modular Drives
Catalog # MFLX77300-01
undefined
Masterflex® L/S® Gear Service Kits for 10 to 600 rpm Digital Modular Drives
Catalog # MFLX77300-01
About this item
Make the most of your Masterflex drives replacement parts and service kits.
  • Easy to install, these service kits avoid costly downtime of your gear pump head
  • Suitable for use with drives 77301-20 and -30.
Extend the life of your Masterflex pump with these replacement parts.
Delivery: Includes gear assembly (nylon gear/shaft), and gear case cover gasket.
  • Description:
    Gear Service Kit for Modular Drives 77301-20/-30
  • Environmentally Preferable:
  • Warranty:
    2 years
  • Compatible with:
    77301-20 or 77301-30
  • Pk:
    1
Frequently Bought Together