Order Entry
Austria
ContactUsLinkComponent
 
Masterflex® Single-Use Quattroflow™ Disposable Pump Chamber with EZ-Set, Polypropylene, 3/4" Tri-Clamp
  MFLX76000-03
undefined
Masterflex® Single-Use Quattroflow™ Disposable Pump Chamber with EZ-Set, Polypropylene, 3/4" Tri-Clamp
  MFLX76000-03

 

Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.

  • EZ-Set design allows for quick pump head change—reducing downtime
  • Constructed with a disposable polypropylene chamber—ideal for single-use applications
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid

The Masterflex® Quattroflow™ single-use replacement pump heads offer a unique four diaphragm (chamber) design, providing the constant low-pulsation, low-shear pumping needed for your biological fluids. In addition, the quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.

Single-use pump heads have a disposable wetted chamber constructed of solid polypropylene (PP) that can be replaced as a complete unit. This saves you time and money by eliminating the cleaning validation/sterilization process. Plus, fluid path integrity is ensured with no chance of cross-batch or cross-product contamination. These single-use pump heads can be sterilized via autoclave or gamma irradiation. With the EZ-Set pump head design, you can change your single-use pump chamber in 30 seconds without the use of special tools or torque wrenches.

Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.

  • Maximum Flow Rate:
    14 L/min
  • Max. pressure:
    58 psi
  • Description:
    Disposable Pump Chamber for Pump System 74000-59
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    76000-03
  • Wetted materials:
    Polypropylene (housing, valve plate), TPE (diaphragm), EPDM(valves, O-rings)
  • Inlet connection:
    3⁄4" Tri-Clamp
  • Outlet connection:
    3⁄4" Tri-Clamp
  • Max. temperature:
    60 °C (140 °F)
  • Series:
    Quattroflow
  • Pk:
    1
Frequently Bought Together