ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Elbow, Female Luer to Female Luer Adapter; 25/PK
Masterflex® Fitting, Polypropylene, Elbow, Female Luer to Female Luer Adapter; 25/PK
Catalog # MFLX45508-26
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Elbow, Female Luer to Female Luer Adapter; 25/PK
Catalog # MFLX45508-26
CE certificate
About this item
Full material traceability to ensure process integrity.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100000 environment
  • Connection Type: Female Luer to Female Luer
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Connection Type:
    Female Luer Union
  • Flow pattern:
    Elbow
  • Description:
    Fitting, Polypropylene, Elbow, Female Luer to Female Luer Adapter; 25/PK
  • Seq:
    01
  • Environmentally Preferable:
  • Model:
    45508-26
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Female luer
  • No. of ports:
    2
  • Pk:
    25
  • Fitting material:
    Polypropylene
Frequently Bought Together