ContactUsLinkComponent
Masterflex® Fitting, Stainless Steel, Luer Stopcock, Female Luer x Male Luer
Masterflex® Fitting, Stainless Steel, Luer Stopcock, Female Luer x Male Luer
Catalog # MFLX41607-00
undefined
Masterflex® Fitting, Stainless Steel, Luer Stopcock, Female Luer x Male Luer
Catalog # MFLX41607-00
  • Connection type (port 1):
  • Female luer
  • Female luer
  • Connection type (port 2):
  • Male luer lock
  • Female luer
  • Connection type (port 3):
  • Male luer lock
  • No. of ports:
  • 2
  • 3
  • Currently Viewing
Specifications
About this item
Exceptional durability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
  • Full material traceability on all items
Easily connect your tubing to your system with these durable luer stopcocks.
  • Description:
    Stainless steel stopcocks Female luer x male luer. 3 months warranty
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Male luer lock
  • No. of ports:
    2
  • Pk:
    1
Frequently Bought Together