Masterflex® Fitting, Black PVDF, One-Way, Stopcock with Female Luer Lock Connectors; 10/PK
MFLX31200-92
- Model:
- 31200-92
- 31200-90
- 31200-91
- Wetted materials:
- Black PVDF
- Natural PVDF, Polypropylene
- Black PVDF
- Black PVDF
Build leakproof fluid handling systems.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
- Flow pattern:One-Way
- Description:Luer stopcock, 1-way, female luer x female luer, black PVDF, pack of 10
- Seq:04
- Environmentally Preferable:
- Model:31200-92
- Wetted materials:Black PVDF
- Pk:10
- Inlet port:Female Luer
- Outlet port:Female Luer
Frequently Bought Together