ContactUsLinkComponent
Masterflex® Fitting, Polycarbonate, Two Ports, Manifolds with Female Luer Locks, 180° Rotation, Non-Sterile; 10/PK
Masterflex® Fitting, Polycarbonate, Two Ports, Manifolds with Female Luer Locks, 180° Rotation, Non-Sterile; 10/PK
Catalog # MFLX30600-44
undefined
Masterflex® Fitting, Polycarbonate, Two Ports, Manifolds with Female Luer Locks, 180° Rotation, Non-Sterile; 10/PK
Catalog # MFLX30600-44
  • Wetted materials:
  • Polycarbonate
  • Polycarbonate
  • Polycarbonate
  • Polycarbonate
  • Warranty:
  • Out of box
  • Out of box
  • Out of box
  • Currently Viewing
Specifications
About this item
Regulate flow without any leaking.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
  • Flow pattern:
    2 Ports, 180 Degree Rotation
  • Description:
    Fitting, Polycarbonate, Two Ports, Manifolds with Female Luer Locks, 180° Rotation, Non-Sterile; 10/PK
  • Environmentally Preferable:
  • Wetted materials:
    Polycarbonate
  • Pk:
    10
  • Inlet port:
    Female Luer
  • Outlet port:
    Female Luer
Frequently Bought Together