ContactUsLinkComponent
Masterflex® Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 48" Long; 5/PK
Masterflex® Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 48" Long; 5/PK
Catalog # MFLX30526-18
undefined
Masterflex® Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 48" Long; 5/PK
Catalog # MFLX30526-18
About this item
Get a seamless transition from source to system.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: female luer x male luer lock with 48" long tubing
This high pressure flexible PVC tubing has luer fittings at each end and comes in various lengths. Can be sterilised by ETO.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
  • Connection Type:
    Tubing Assembly
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 48" Long; 10/PK
  • Seq:
    03
  • Environmentally Preferable:
  • Model:
    30526-18
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Male luer lock
  • No. of ports:
    2
  • Warranty:
    Out of box
  • Pk:
    10
  • Fitting material:
    Polycarbonate
Frequently Bought Together