Order Entry
Austria
ContactUsLinkComponent
PendoTech PDKT-650-298SC Single-Use Cable with RJ12 to Scilog Pump; 6' (1.8 m)
PendoTech PDKT-650-298SC Single-Use Cable with RJ12 to Scilog Pump; 6' (1.8 m)
Catalog # MFLX19406-85
CE Compliant
undefined
PendoTech PDKT-650-298SC Single-Use Cable with RJ12 to Scilog Pump; 6' (1.8 m)
Catalog # MFLX19406-85
CE Compliant

About this item

Optimize performance of your PendoTECH® Pressure Sensor.

  • Choose from a wide range of replacement parts and accessories

Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.

  • Length:
    182,88 cm (72")
  • Description:
    Cable with RJ12 to Scilog Pump, 6 ft (1,8 m)
  • Environmentally Preferable:
  • Warranty:
    2 years
  • Pk:
    1
Frequently Bought Together