PendoTech TT1 Single-Use Temperature Sensor Transmitter with 4-20 mA Output
Catalog # MFLX19406-45
Supplier: Avantor Fluid Handling
2 year warranty
About this item
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
- Produces a 4-20 milliamp signal that is linear with temperature
- Connections to the transmitter are quick and convenient
- Convenient mounting screws
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
The temperature sensor transmitter easily connects to the PendoTECH temperature sensor. The narrow 4-20mA output range from 0 to 70ºC ensures accurate and drift free linearization of the sensor output.
- Description:Temperature Sensor Transmitter with 4-20mA Output
- Environmentally Preferable:
- Warranty:2 years
- Pk:1
Frequently Bought Together