PendoTech Single-Use Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
Catalog # MFLX19406-34
Supplier: Avantor Fluid Handling
About this item
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Connects flow through tubing to luer fitting single use pressure sensors.
- Max. pressure:76 psi
- Process connection:1/4" x 1/4" hose barb, male luer
- Description:Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
- Environmentally Preferable:
- Warranty:3 Months
- Pk:1
Frequently Bought Together