PendoTech Single-Use Temperature Sensor Cable with 1/4" Phone Jack for Hosebarb Sensors; 10 ft (3 m)
MFLX19406-15
CE Compliant
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
- Length:304,8 cm (120")
- Description:Reusable Temperature Sensor Cable with 1/4" Phone Jack for Hose Barb Fitting; 10 ft (3 m)
- Environmentally Preferable:
- Warranty:2 years
- Pk:1
Frequently Bought Together