Order Entry
Austria
ContactUsLinkComponent
Masterflex® Ismatec® Footswitch for Digital Drives, 3-pin circular connector; 6-ft (1,8 m)
Masterflex® Ismatec® Footswitch for Digital Drives, 3-pin circular connector; 6-ft (1,8 m)
Catalog # MFLX07610-25
undefined
Masterflex® Ismatec® Footswitch for Digital Drives, 3-pin circular connector; 6-ft (1,8 m)
Catalog # MFLX07610-25

About this item

Simplify your dispensing.

  • Select the footswitch with connectio suitable for your Ismatec® pump

Footswitches provide convenient, hands-free operation of your pump drives from up to six feet away. Simply press once to start pump, press again to stop.

  • Description:
    Footswitch for Digital Drives, 3-pin circular connector; 6-ft (1,8 m)
  • Length:
    183 cm
  • Seq:
    01
  • Environmentally Preferable:
  • Model:
    07610-25
  • Input:
    3-pin circular connector
  • Warranty:
    1 year
  • Series:
    Ismatec Digital Drives
  • Pk:
    1
Frequently Bought Together