ContactUsLinkComponent
Masterflex® L/S® Peristaltic Pump Drive Replacement Brushes; 2/PK
Masterflex® L/S® Peristaltic Pump Drive Replacement Brushes; 2/PK
Catalog # MFLX07533-01
undefined
Masterflex® L/S® Peristaltic Pump Drive Replacement Brushes; 2/PK
Catalog # MFLX07533-01
About this item
Quickly and easily replace components of your Masterflex system.
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
  • Description:
    L/S® Replacement Brushes, 2/PK
  • Environmentally Preferable:
  • Pk:
    2
Frequently Bought Together