ContactUsLinkComponent
Masterflex® Replacement Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
Masterflex® Replacement Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
Catalog # MFLX07528-82
undefined
Masterflex® Replacement Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
Catalog # MFLX07528-82
About this item
Make the most of your Masterflex® Open-Head Sensor with these accessories and replacement parts.
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.
  • Description:
    Replacement Masterflex Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
  • Environmentally Preferable:
  • Warranty:
    2 years
  • Pk:
    1
Frequently Bought Together